Anti-NDUFB7 Mouse Monoclonal Antibody
Leverandør: Abgent
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABGEAT3005AEA 2620 DKK
ABGEAT3005A
Anti-NDUFB7 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: NDUFB7 (NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7, 18kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human