Anti-NDOR1 Mouse Monoclonal Antibody
Leverandør: Abgent
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABGEAT2990AEA 2620 DKK
ABGEAT2990A
Anti-NDOR1 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: NDOR1 (NADPH dependent diflavin oxidoreductase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human