Anti-MAD2L1 Mouse Monoclonal Antibody

Leverandør: Abgent
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABGEAT2758AEA 2620 DKK
ABGEAT2758A
Anti-MAD2L1 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: MAD2L1 (MAD2 mitotic arrest deficient-like 1 (yeast))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR