Anti-C1orf177 Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AVIVARP53062_P050EA 2770 DKK
AVIVARP53062_P050
Anti-C1orf177 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: C1orf177 (chromosome 1 open reading frame 177)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR