Anti-BBX Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AVIVARP39409_P050EA 2770 DKK
AVIVARP39409_P050
Anti-BBX Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BBX (bobby sox homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR