Anti-TEF Rabbit Polyclonal Antibody

Leverandør: Aviva Systems Biology
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AVIVARP37237_T100EA 2410 DKK
AVIVARP37237_T100
Anti-TEF Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TEF (thyrotrophic embryonic factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR