Blood stain card, Whatman™
Leverandør: Qiagen
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Whatman™
512-1150EA 2108.84 DKK
512-1150
Blood stain card, Whatman™
Opbevaringssystemer til DNA og RNA
For the collection and transport of blood and body fluids.
  • Made from absorbent filter paper
  • Enables efficient sample collection
  • Features a protective cover that aids sample drying and protects sample from direct contact
  • Designed for short-term handling of specimens