Vacuum pressure indicator for vacuum desiccators, SP Bel-Art
Leverandør: Bel-Art
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BELAH420420000EA 728 DKK
BELAH420420000
Vacuum pressure indicator for vacuum desiccators, SP Bel-Art
Vacuum håndteringssystemer
The plastic bellows collapse under negative pressure vacuum within the desiccator and expand under ambient atmospheric pressure. Fits easily onto the tapered end of the stopcock.