School fume hoods, benchtop, SP Bel-Art
Leverandør: Bel-Art
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BELAH500002002EA 18400 DKK
BELAH500002002
School fume hoods, benchtop, SP Bel-Art
Stinkskabe og kabinetter Aftræksskabe til apoteker
Fume hood provide maximum visualization and protection during lessons. The front sash window opens to three adjustable positions for air flow control. An exhaust system in the hood capable of drawing 200 cfm (free air).
  • Light weight
  • Chemical resistant
  • Easy to clean
Construction: Polyethylene hood, polycarbonate windows
Shipping weight: 18,1 kg