Disinfection trays
Leverandør: Phagogene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PHAG1077EA 3890 DKK
PHAG1077 PHAG1079 PHAG1078
Disinfection trays
Bakker/Tallerkener/Potter Sterilisationsbakker/-skåle
Trays used for pre-disinfection and disinfection of surgical instruments.
  • Hollow handles for easy grip
  • Transparent cover for ease of use
  • Graduation at the interior of the trays