VWR® Catalase Reagent
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
941970ZAEA 246 DKK
941970ZA
VWR® Catalase Reagent
Mikrobiologiske testsystemer Enzymatiske Test
The catalase test can differentiate or identify microorganisms by checking their ability to produce catalase, an enzyme that protects microorganisms from toxic oxygen metabolites. Usually only aerobic microorganisms produce catalase.
  • A simple test to identify aerotolerant strains
  • Allows identification of Gram-positive cocci
  • Easy to use
  • Storage: 8 to 25 °C