Multi-function air quality measuring apparatus, testo 480
Leverandør: Testo
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TEST0563.4800EA 12100 DKK
TEST0563.4800
Multi-function air quality measuring apparatus, testo 480
This measuring instrument is designed to analyse the ventilation and indoor air quality of the working environment, serving as an indicator for the wellbeing of people at their workplaces and as a monitor in storage and production process areas.
  • Indoor air quality alerts whether the air conditioning system (HVAC) is working to maximum efficiency
  • Absolute pressure, draught, Lux, U-value, CO₂, temperature and volume flow can additionally be determined
* Differential pressure
** Absolute pressure
Leveringsinformation: Supplied with a PC software, and USB cable.