Liquid nitrogen cryostorage systems, CryoPlus™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
THBR7401EA 128750 DKK
THBR7401 THBR7405-MAR
Liquid nitrogen cryostorage systems, CryoPlus™
The ideal combination of liquid nitrogen, storage reliability and microprocessor technology, the CryoPlus Series LN₂ Storage system provides precise and accurate control over all parameters without complicated programming.
  • Low-profile tank with vacuum-insulated
  • Stores up to 5500 to 6318 vials (2 ml)
  • Alarm contacts for remote monitoring of storage tank conditions
  • Precise liquid level control with over 16 parameters with autofill system
  • Two independent, flexible lid gaskets reduce moisture migration into the chamber