Erlenmeyer flasks with vent cap
Leverandør: Corning®
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Corning®
734-2759EA 4170 DKK
734-2759 734-2758
Erlenmeyer flasks with vent cap
Kolber Dyrkningsbeholdere
Polycarbonate, transparent.
  • Moulded-in graduations for accuracy
  • Highest Sterility Assurance Level (SAL)
  • Certified non pyrogenic and free from RNase and DNase
  • Erlenmeyer flasks are autoclavable once at 121 °C, 15 psi for 20 minutes
  • Repeated autoclaving weakens the flask and is not recommended
  • Vent cap is not autoclavable
This Erlenmeyer flask is designed to help scale-up suspension culture with space efficiency in mind. The flasks are sterile and ready-to-use, ideal for shaker culture applications and storage. Plain or baffled bottom. Large vent cap for continuous gas exchange while ensuring sterility and preventing leakage.