Temperature and humidity loggers
Leverandør: Dickson
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DICKTP425EA 11490 DKK
DICKTP425
Temperature and humidity loggers
Dataloggere
The data logggers provides with current minimum and maximum data is displayed on the easy-to-read LCD screen.
  • Display flashes when the logger is full
  • Logger starts and stops with the push of a button
  • Features high speed USB connectivity and a user replaceable battery
Display resolution: 0 - 999
Display update rate: 20 sec
Certificeringer: IP 20 certified protection class.
Leveringsinformation: Product comes with 1 year of warranty.