Flow coolers, Techne®
Leverandør: Cole Parmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TECNFFC2DEA 34720 DKK
TECNFFC2D TECNFFC5D
Flow coolers, Techne®
Techne® flow coolers work in conjunction with a Techne® thermoregulator. The bath liquid flows through the unit which continually extracts heat from the bath fluid by means of the heat exchanger which is built into the unit.
  • Compact refrigeration units for achieving temperatures down to –35 °C*
  • Designed for use with liquid baths
Advarsel: Cooling capacity at +20, 0 and −10 °C.
* At an ambient of 20°C, using a mixture of 40% water, 40% antifreeze and 20% ethanol