Sample Preparation Racks, Thermo Scientific
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ADVAPRK-1383EA 2840 DKK
ADVAPRK-1383
Sample Preparation Racks, Thermo Scientific
The 384-well aluminium support rack provides optimal support for heat sealing and automated dispensing for all standard 384-well PCR plates as well as 96-well Piko PCR plates and plate/frame assemblies.
  • V-shaped wells give optimal support for ultra-thin walls for applying and removing sealers
  • Industry standard footprint fits in robotic workstations, enabling automated pipetting with Piko PCR plates
  • For use with real-time PCR systems