Capillary viscometers, Cannon-Fenske Opaque
Leverandør: Fungilab
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FUNGCV004-103EA 820 DKK
FUNGCV004-103 FUNGCV004-104 FUNGCV004-105 FUNGCV004-106 FUNGCV004-107 FUNGCV004-108 FUNGCV004-109 FUNGCV004-110 FUNGCV004-111 FUNGCV004-101 FUNGCV004-112 FUNGCV004-102
Capillary viscometers, Cannon-Fenske Opaque
Viskosimetre Kapillærviskosimetere
Cannon-Fenske Opaque viscometers (reverse flow type) for dark Newtonian liquids especially for liquids so dark in colour that cannot be seen in a Cannon-Fenske routine viscometer.
Used to study shearing stress and shearing rate.
Certificeringer: ASTM D-445-446, ASTM D2515, ISO 3104-3105 and UNE 400313
Leveringsinformation: Supplied with the calibration certificate granted by a National Metrological Register with recognised international standards UKAS.