Hand and surface sanitising wipes, KLEENEX®
Leverandør: Kimberly-Clark
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-1736EA 90.8 DKK
115-1736
Hand and surface sanitising wipes, KLEENEX®
Papirservietter Engangsservietter og - håndklæder Desinficerende servietter
These non alcohol wipes are ideal for sanitising hands, desks and work stations, and other surfaces. The wipes effectively kills up to 99,999% of common bacteria.
  • Ideal for applications where a product containing alcohol can not be used
  • Instant results in as little as 30 seconds
  • Fragrance and dye free
  • Dispensed from a centrefeed tub dispenser: Ideal for communal areas such as meeting, conference rooms and break room areas
Advarsel: Use biocides safely. Always read the label and product information before use.