VWR® Plugs for Drosophila Vials and Bottles
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
734-2875EA 3150 DKK
734-2875 734-2672 734-2673
VWR® Plugs for Drosophila Vials and Bottles
Propper Cellulosepropper
These plugs are made of bonded dense weave cellulose acetate which provides an excellent barrier to mite infestation. They are precision manufactured for consistent fit and minimal waste.
  • Reduced evaporation, which keeps food fresh longer
  • Minimal shedding compared to cotton and rayon
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904