Protease Estimation Assay, ProteSEEKER™, G-Biosciences
Leverandør: G-Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ProteSEEKER™
786-325EA 8230 DKK
786-325
Protease Estimation Assay, ProteSEEKER™, G-Biosciences
Assays Enzym Assays
ProteSEEKER™ protease screening assay consists of a ready-to-use dye-labeled protein, which is digested by proteases to release dye-labeled peptides. The absorbance of which is measured for determination of protease activity. The inhibitors are supplied at a 100X concentration and the 1X concentration provides >90% inhibition in most biological samples.
Bestillingsoplysninger: Accessories must be ordered separately.