VWR® Measuring Beakers with Handle, Porcelain
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
213-0409EA 619 DKK
213-0409 213-0408 213-0412 213-0411 213-0410
VWR® Measuring Beakers with Handle, Porcelain
Bægre
Porcelain, glazed white.
  • Exceptional chemical resistance
  • Excellent thermal resistance
  • Robust, heavy construction
Cylindrical, with spout. Very easy to read, internal graduations.