Vil du logge ind igen?
OBS! Det er ikke længere muligt at skrive til @dk.vwr.com benyt istedet @vwr.com. Kontakt evt vores kundeservice.
UltraMOP™III cleanroom wall mop heads
Leverandør: FOAMTEC INTERNATIONAL, LLC
UltraMOP™
UltraMOP™ III mop heads are constructed of a snag-resistant, polyester fabric laminated to an absorbent polyurethane foam employing the patent pending Bump-PROOF® design. The Bump-PROOF® design improves cleanliness and durability as the durable fabric extends over the front and rear edges making it ideal for ISO class 4 and 5 clean rooms.
  • Compatible with gamma, autoclave and EtO sterilization methods
  • Constructed of a proprietary, super, fine pore polyurethane foam that is ideal to laydown a precise volume of disinfectant cleaner
  • Laminated fabric covers the leading and trailing edge preventing abrasion related particle contamination
  • Product traceability: available with lot Certification of Conformance to ease audit and validation procedures
  • Simple to use: bump-proof wrap-around design eliminates the need for protective polyester sock
  • Robust cleaning: microfiber versions remove stubborn contamination and are excellent where dry floors are desired
BESTIL
Udvid alle / Skjul alle
Beskrivelse Farve Længde Pakn VWR katalog nummer Mængde
UltraMOP™III cleanroom wall mop head, polyester lamination, sterile blå/hvid 178 mm 32 FOAMFS701ST
UltraMOP™III cleanroom wall mop head, polyester lamination, sterile blå/hvid 355 mm 32 FOAMFS702ST
UltraMOP™III cleanroom wall mop head, wrapped with microfiber lamination w/modified “T” Sterile blå/hvid - 32 FOAMFS703ENST
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Skriv en bedømmelse
Brug for hjælp?
  • Uanset hvad du mener, positivt eller negativt, er det vigtigt for os.
  • Det tager kun få sekunder at give os feedback.
  • Din feedback vil blive offentliggjort på internettet.
VWR.com vil offentliggøre dit navn som vwr.com bruger.
Du kan bruge dette navn, eller redigere og gemme et kaldenavn.
Vi anbefaler at brugerne bruger deres første og sidste initialer
Bekræft venligst branchen du arbejder i
Eksempel: farmaceutisk
Bekræft venligst dit arbejde
Eksempel: Klinisk assistent
Bekræft venligst Byen
Eksempel: New York
Bedømmelser
Samlet vurdering
Gennemsntilig vurdering
Sortér alle efter:
Enter Details
Select the article for which you are looking for Certificate
Anfør Lotnummeret. Feltet er mandatory. skriv "x" for VWR kemikalier hvis du ikke kender nummeret
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse katalog Nummer Availability Enhed Din pris Price Per Antal