Single-use data loggers, Testo 184 T1 and Testo 184 T2
Leverandør: Testo
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-2306EA 400 DKK
620-2306 620-2309
Single-use data loggers, Testo 184 T1 and Testo 184 T2
Dataloggere
These data loggers are designed for an accurate monitoring, avoiding interuption of the cold chain process in quality control. They travel in freight and loading rooms to monitor temperatures of sensitive goods.
  • Clear indication: Red/green LED alarm
  • Easy and intuitive mode of use, quick configuration
  • Clear PDF readout and reporting
  • Secure IT integration without any installation
  • Connect the logger to a PC and produce a PDF report
  • Warranty: 12-month
In order to work more efficiently, with the dataloggers all the files are stored directly and securely in the respective testo 184. No software installation required on PC or notebook in order to configure a testo 184 data logger. The pdf configurationn tool is stored in the logger itself and can be opened with the simple double-click.
If advance read out options are required, the customer can obtain these by purchasing ComSoft Professional.
Certificeringer: EN 12830; CE 2014/30/EU; DIN EN 50581:2013; DIN EN 61326-1:2013; 2011/65/EU standards and RTCA/DO-160G authorisations.