Protective hand creams, Ligana® A Oleo-Tec
Leverandør: Faweco Hautschutz
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Lindesa® O
FAWE12952013EA 231 DKK
FAWE12952013
Protective hand creams, Ligana® A Oleo-Tec
Hudpleje Cremer Hand Creams
Lindesa® O Skin protection cream is especially suitable when working with water-insoluble materials like varnishes, oils, petrol, resins, paints, organic solvents, coating compounds, or similar material, as well as caring for degreased skin after work.
  • Moderately greasy cream
  • Oil in water emulsion
  • Silicone-free, perfumed
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904