Hand disinfectant gels and solutions, Phagorub SPS
Leverandør: Phagogene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Phagorub
PHAG65047EA 1090 DKK
PHAG65047 PHAG65023 PHAG65045 PHAG60672 PHAG60673 PHAG60674
Hand disinfectant gels and solutions, Phagorub SPS
Hudpleje Hånddesinfektion Gel Skin Disinfectants
Hydroalcoholic gels and solutions for the disinfection of the hands. Special formulation enriched with moisturisers and emollients, ideal for sensitive skin.
  • Bactericidal, mycobactericidal, fungicide, virucidal
  • Ethanol based
  • Gel viscosity suitable for hygienic controlled friction: 3 ml for 15 seconds
  • Fragrance-free dyes
Advarsel: Use biocides safely. Always read the label and product information before use.