Viscosity measurement control units
Leverandør: Lauda
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LAUDLMV812EA 54850 DKK
LAUDLMV812 LAUDLMV813
Viscosity measurement control units
The PVS control unit is key to the PVS automated systems.
It is also the switching station between the PC and individual components such as the S5 stand or the VRM cleaning modules or the MT dosing systems.