Calcium pantothenate
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A2088.0025EA
0
DKK
A2088.0025
Calcium pantothenate
Calcium pantothenate
Formel:
C₁₈H₃₂CaN₂O₁₀ Molekylvægt: 476,54 g/mol |
MDL Number:
MFCD00002766 CAS-nummer: 137-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...