Vial identification system, Virtuoso™
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CRMA60180-VT100EA 79470 DKK
CRMA60180-VT100
Vial identification system, Virtuoso™
From sample entry to final reports, everything an analytical lab does revolves around ensuring accurate data and reliable results. Chromatography vial sample identification can be complicated, because once the sample is in the vial, there is no easy way to identify it. Current methods, like hand written or adhesive labels, can be illegible or time consuming. The Virtuoso™ Vial Identification System is the most innovative device for ensuring sample identity and sample security. It is a fast, accurate, detailed and reproducible system for producing customised sample information directly onto a vial.
  • Load up to 10 vials with the included multi-vial sleeve
  • Add text, graphics, barcodes and logos onto the vials
  • Input data instantly from the touch screen interface
  • Network connectivity allows you to transfer label information from your files