Du Søgte efter: Electricity/Magnetism Learning Activities
40  results were found
SearchResultCount:"40"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Varenummer: (765-0311)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: Aluminum wall rail with 20 removable plastic brackets. Mounted on the wall. Screw included. Dimensions: rail length 500 mm, height 40 mm. console leng...
UOM: 1 * 1 Items
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: Learn the principles of magnetism with these two activity sets designed for middle and high school students. Two sets of activities each with 10 exper...
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: Electronic buzzers for a DC supply.
Varenummer: (761-0163)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: Educational Materials, Electricity/Magnetism, Magnetic track
UOM: 1 * 1 Items
Varenummer: (766-0277)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: Very affordable basic set with solid components. Contacts of neodymium magnets for easy connection to battery and accessories.
UOM: 1 * 1 Items
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: 60 sorted, fast fine fuses. 10 pcs of each type. In compartmentalized plastic box with transparent lid. Fuse dimensions: 5×20 mm.
Varenummer: (766-0300)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: For pulleys and gears with 4 mm shaft holes when they are to be mounted on a 4 mm motor shaft. Pack of 20 pcs.
UOM: 1 * 20 Items
Varenummer: (761-0158)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: WindPitch is a miniature real-working wind turbine (wind power generator) designed for students to evaluate the pitch (setting angle) of the profiled ...
UOM: 1 * 1 Items
Varenummer: (765-0284)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: Experimental set in Ellära and Magnetism, primarily intended for compulsory school upper secondary school or equivalent level of knowledge. The experi...
UOM: 1 * 1 Items
Varenummer: (765-0597)
Leverandør: ELWE LEHRGERAETEBAU KLINGENTHA
Beskrivelse: The exercise oscilloscope is placed at the Helmholtz coils so that its main axis coincides with the axis of the coils. A sawtooth-shaped voltage is ap...
UOM: 1 * 1 Items
Leverandør: GLASWARENFABRIK KARL HECHT
Beskrivelse: According to Hoffmann, with two city dishes, 0 to 30 ml of grading graded 1/10 ml.
Leverandør: CORNELSEN EXPERIMENTA GMBH & C
Beskrivelse: Bare wire in a roll, on a plastic former.
Varenummer: (765-0198)
Leverandør: CORNELSEN EXPERIMENTA GMBH & C
Beskrivelse: Insulated rod with 3 connection terminals for wire up to 2 mm and 4 mm contact sleeve.
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Varenummer: (765-0201)
Leverandør: CORNELSEN EXPERIMENTA GMBH & C
Beskrivelse: Used in electrochemistry. Voltage, max 2 V. The unit is divisible and consists of glass beaker, cylindrical zinc electrode, plastic beaker, carbon ele...
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Varenummer: (765-0112)
Leverandør: CORNELSEN EXPERIMENTA GMBH & C
Beskrivelse: Plastic rod with metal tube equipped with an axial and a radial 4 mm connection.
UOM: 1 * 1 Each
Market Source Item This is a MarketSource item. Additional charges may apply
Varenummer: (765-0109)
Leverandør: CORNELSEN EXPERIMENTA GMBH & C
Beskrivelse: The kit contains about 15 different components with which a number of experiments can be performed. The components are designed so that students can q...
UOM: 1 * 1 Each
Market Source Item This is a MarketSource item. Additional charges may apply
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 40
no targeter for Bottom