Du Søgte efter: Fysik undervisningsmateriale
141  results were found
SearchResultCount:"141"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Varenummer: (201-0216)
Leverandør: GLASWARENFABRIK KARL HECHT
Beskrivelse: Five communicating glass tubes with different inner diameters. Used to show capillary forces and how they are affected by the inner diameter of the pi...
UOM: 1 * 1 Items
Varenummer: (765-0969)
Leverandør: GLASWARENFABRIK KARL HECHT
Beskrivelse: Five glass tubes of different shape and inner dimensions. Down to each other. When filling the liquid in the pipes, it is clear that the height of the...
UOM: 1 * 1 Items
Leverandør: DIDALAB
Beskrivelse: Educational Materials, Optics, Applikation: Physics, Diode laser, blue, Class II
Leverandør: DIDALAB
Beskrivelse: This comprehensive optical pack allows you to implement and study the basic principles of polarisation of light.
Leverandør: DIDALAB
Beskrivelse: These wide diameter lenses are used for easy classroom experiments and for aberrations underscoring.
Leverandør: DIDALAB
Beskrivelse: Educational Materials, Optics, Applikation: Physics, Line generator, with a laser, enables to project one line, by the use of one glass cylinder, rotating at 360°
Leverandør: DIDALAB
Beskrivelse: Educational Materials, Optics, Applikation: Physics, Translucent screen 20×20 cm, attached to a rod with Ø 10 mm, one graduated side
Varenummer: (DIDAPOD060056)
Leverandør: DIDALAB
Beskrivelse: Triangular, cast aluminium optical bench
UOM: 1 * 1 Items
Varenummer: (DIDAPOD013135)
Leverandør: DIDALAB
Beskrivelse: The diameter of the beam is adjustable from 1 to 12 mm.
UOM: 1 * 1 Items
Varenummer: (DIDAPOD013132)
Leverandør: DIDALAB
Beskrivelse: Extremely luminous beam, ideal for classroom experiments. Class IIIa (5 mW Power)
UOM: 1 * 1 Items
Varenummer: (DIDAPOD010240)
Leverandør: DIDALAB
Beskrivelse: Optical glass components are manufactured with high accuracy polishing and flatness tolerance.
UOM: 1 * 1 Items
Varenummer: (DIDAPOD010615)
Leverandør: DIDALAB
Beskrivelse: Prism made of standard optical glass with index near to 1,52. Equilateral dispersing prisms are used for wavelength separation applications. Intended ...
UOM: 1 * 1 Items
Leverandør: DIDALAB
Beskrivelse: High quality professional gas laser, Class II
Varenummer: (216-0428)
Leverandør: Thermo Fisher Scientific
Beskrivelse: Replace metal-type troughs with the polypropylene pneumatic trough.
UOM: 1 * 1 Items
Leverandør: DIDALAB
Beskrivelse: Economical, recent design of prismatic optical bench
Varenummer: (765-0311)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: Aluminum wall rail with 20 removable plastic brackets. Mounted on the wall. Screw included. Dimensions: rail length 500 mm, height 40 mm. console leng...
UOM: 1 * 1 Items
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 141
no targeter for Bottom