Sharp disposal container Multi-Safe quick
Leverandør: Sarstedt
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SARS77.3891.010EA 1840 DKK
SARS77.3891.010
Sharp disposal container Multi-Safe quick
Boxes, made of PP, yellow/red. The practical safety box for user-friendly disposal of sharp, pointed and dangerous objects in the stationary and ambulatory fields.
All boxes can be attached to any flat surface/tray using a adhesive adapter to prevent them from overturning (see accessories).
Certificeringer: TRBA 250, DIN EN ISO 23907