B.O.D. measurement, BOD EVO Sensor System 6
Leverandør: VELP Scientifica
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
705-0780EA 16680 DKK
VELPS10220156 VELPS10220146
B.O.D. measurement, BOD EVO Sensor System 6
Respirometric measurement system for BOD without mercury for determining the amount of dissolved oxygen consumed by aerobic biological microorganisms in water.
  • Very simple to use and mercury-free
  • Real time data (in ppm BOD) is displayed on the sensor head of each bottle
  • Using wireless data transmission five-day test can record results every 30 min up to every 24 H
  • Six sample bottles with a magnetic stirring station for ensuring reproducibility
  • Measurement ranges: 90/250/600/999 ppm BOD
The sensor uses manometric technology and microprocessor control to provide regular data measurements over defined periods. The sensors require two lithium batteries each and are IP 54 rated. Using the Wireless Databox™ and BODSoft™, the data from up to 80 samples can be monitored, compared and and stored.
Temperature control is usually required for the most reproducible results - please see entries for thermostatic cabinets and incubators which are supplied separately.
Leveringsinformation: The BOD EVO Sensor systems include a six-position stirring station with BOD sensors, dark glass sample bottles, alkali holders and stirring bars.