Homogeniser, OV5
Leverandør: VELP Scientifica
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VELPR20900010EA 7290 DKK
VELPR20900010
Homogeniser, OV5
Stand model, volume range 0,1 ml to 8 L. Homogeniser for dispersing, homogenising and mixing biological tissues (cells, animal and vegetable tissues), pharmaceutical products, cosmetics and food products. The ergonomic design allows the user to use one shaft with different rotors and stators according to the user application.
  • One shaft for all applications with selection of rotors and stators to match the application
  • Stirring volume up to 8 L (water) as homogeniser
  • Stirring volume up to 40 L (water) as high speed mixer
  • Handles fluids up to 10000 mPa.s (cP)
  • Speed variable from 10000 to 30000 min⁻¹ (rpm)
  • One shaft for all applications with selection of rotors and stators to match the application
  • Stirring volume up to 8 litres (water) as homogenizer
  • Stirring volume up to 40 litres (water) as high speed mixer
  • Handles fluids up to 10 000 mPa.s (cP)
  • Speed variable from 10 000 to 30 000 min-1 (rpm)
Homogeniser for dispersing, homogenizing and mixing biological tissues (cells, animal and vegetable tissues), pharmaceutical products, cosmetics and food products. The ergonomic design allows the user to use one shaft with different rotors and stators according to the user application.
Bestillingsoplysninger: Stand and clamps and appropriate rotor/stator need to be ordered separately.