Reagent reservoir
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BHIT783500EA 479 DKK
BHIT783500
Reagent reservoir
Reagens reservoirs
PP, white. This durable reagent vessel is chemically resistant to all common reagents. It is especially useful with all multi channel pipettes.
  • Chemically resistant
  • Autoclavable