Dry ice maker, Frigimat®, SP Bel-Art
Leverandør: Bel-Art
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SP Bel-Art Frigimat®
BELAH388780012EA 27920 DKK
BELAH388780012
Dry ice maker, Frigimat®, SP Bel-Art
Tøris maskine
Dry ice maker made of epoxy-coated steel with an aluminum ice chamber and brass valves with four non-skid rubber feet. Includes a 1,8 m nickel plated copper connecting tube with standard CGA 320 (0.830"-14) thread couplings for easy attachment to an LCO₂ cylinder.
  • It employs the Joule- Thomson effect to produce a solid 454 g block of dry ice in one to two minutes, and the block can be removed in just 30 s
  • A brass relief valve safely vents excess gas
  • Each 22,5 kg cylinder of liquid CO₂ with a dip tube will make 8 or 9 blocks of dry ice, each measuring 85×110 mm long.
W×D×H: 300×170×130 mm
Shipping weight: 5,5 kg