5-Bromo-2-hydroxy-6-picoline ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAB2796-1GEA
501
DKK
TCIAB2796-1G
5-Bromo-2-hydroxy-6-picoline ≥98.0% (by GC)
5-Bromo-2-hydroxy-6-picoline
Formel:
C₆H₆BrNO Molekylvægt: 188,02 g/mol |
MDL Number:
MFCD03427653 CAS-nummer: 54923-31-8 |
Specifikationer Testresultater
Purity (GC) | min. 98.0 % |
Melting point | 200.0 to 204.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...