2-Methyl-5-(trifluoromethoxy)benzaldehyde 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H33783.03EA 672 DKK
H33783.03 H33783.06
2-Methyl-5-(trifluoromethoxy)benzaldehyde 96%
2-Methyl-5-(trifluoromethoxy)benzaldehyde
Formel: C₉H₇F₃O₂
Molekylvægt: 204,15 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18399706
CAS-nummer: 195622-40-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR