2-Cyclohexylphenol ≥97.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAC0732-25GEA
1380
DKK
TCIAC0732-25G
TCIAC0732-5G
2-Cyclohexylphenol ≥97.0% (by GC)
2-Cyclohexylphenol
Formel:
C₁₂H₁₆O Kogepunkt: 284°C |
MDL Number:
MFCD00019335 CAS-nummer: 119-42-6 EINECS: 204-322-4 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...