4,4'-Diisopropylbiphenyl

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAD2047-25GEA 591 DKK
TCIAD2047-25G
4,4'-Diisopropylbiphenyl
4,4'-Diisopropylbiphenyl
Formel: C₁₈H₂₂
MDL Number: MFCD00043533
CAS-nummer: 18970-30-4
https://dk.vwr.com/stibo/web/std.lang.all/88/38/28978838.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 68.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR