2-(3,4,5-Trifluorophenyl)ethanol 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H32800.MDEA 328 DKK
H32800.MD H32800.03
2-(3,4,5-Trifluorophenyl)ethanol 96%
2-(3,4,5-Trifluorophenyl)ethanol
Formel: C₈H₇F₃O
Molekylvægt: 176,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06660355
CAS-nummer: 886761-77-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR