Perfluoro(allylbenzene) ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAP1343-5GEA 403 DKK
TCIAP1343-5G
Perfluoro(allylbenzene) ≥98.0% (by GC)
Perfluoro(allylbenzene)
Formel: C₉F₁₀
MDL Number: MFCD00153225
CAS-nummer: 67899-41-6
https://dk.vwr.com/stibo/web/std.lang.all/19/94/28991994.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR