Ammonium perfluorooctanoate
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC15987152EA
2430
DKK
EHERC15987152
Ammonium perfluorooctanoate
Ammonium perfluorooctanoate
Formel:
C₈H₄F₁₅NO₂ |
MDL Number:
MFCD00042599 CAS-nummer: 3825-26-1 EINECS: 223-320-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...