11H-Perfluoroundecanoic acid ≥95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
L16759.14EA
2540
DKK
L16759.14
11H-Perfluoroundecanoic acid ≥95%
11H-Perfluoroundecanoic acid
Formel:
C₁₁H₂F₂₀O₂ Molekylvægt: 546,11 g/mol Smeltepunkt: 102…104 °C Flammepunkt: >100 °C (212 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00042431 CAS-nummer: 1765-48-6 EINECS: 217-184-5 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...