5-Butylpicolinic acid ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAF0227-1GEA 2740 DKK
TCIAF0227-1G
5-Butylpicolinic acid ≥98.0% (by GC)
5-Butylpicolinic acid
Formel: C₁₀H₁₃NO₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00006298
CAS-nummer: 536-69-6
EINECS: 208-643-0
https://dk.vwr.com/stibo/web/std.lang.all/88/30/28968830.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 97.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR