Citronellic acid ≥95.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAC1708-25MLEA
1090
DKK
TCIAC1708-25ML
Citronellic acid ≥95.0% (by titrimetric analysis)
Citronellic acid
Formel:
C₁₀H₁₈O₂ Molekylvægt: 170,25 g/mol |
MDL Number:
MFCD00002728 CAS-nummer: 502-47-6 EINECS: 207-939-7 UN nummer: 2810 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 90.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...