5-(4-Hydroxyphenyl)valeric acid ≥98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B25107.03EA 317 DKK
B25107.03
5-(4-Hydroxyphenyl)valeric acid ≥98%
5-(4-Hydroxyphenyl)valeric acid
Formel: C₁₁H₁₄O₃
Molekylvægt: 194,23 g/mol
Smeltepunkt: 116…120 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04039777
CAS-nummer: 4654-08-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR