2-(4-Hydroxyphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAH1037-25GEA
3440
DKK
TCIAH1037-25G
2-(4-Hydroxyphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
2-(4-Hydroxyphenyl)propionic acid
Formel:
C₉H₁₀O₃ Molekylvægt: 166,18 g/mol Smeltepunkt: 131 °C |
MDL Number:
MFCD00128847 CAS-nummer: 938-96-5 EINECS: 213-352-7 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 129.0 to 133.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...