Octyltin trichloride
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC15715000EA
1190
DKK
EHERC15715000
Octyltin trichloride
Octyltin trichloride
Formel:
C₈H₁₇Cl₃Sn |
CAS-nummer:
3091-25-6 EINECS: 221-435-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...