4-Methyloctanoic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-methylcaprylic acid

TCIAM2048-5GEA 321 DKK
TCIAM2048-5G
4-Methyloctanoic acid ≥98.0% (by titrimetric analysis)
4-Methyloctanoic acid
Formel: C₉H₁₈O₂
Molekylvægt: 158,24
Kogepunkt: 143°C
MDL Number: MFCD00051938
CAS-nummer: 54947-74-9
EINECS: 259-404-2
UN nummer: 3265
ADR: 8,III
https://dk.vwr.com/stibo/web/std.lang.all/79/99/28987999.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR